3-Fluoro-5-formylbenzonitrile ≥95%

Supplier: Matrix Scientific

067013-500MG 067013-1G
101932-968EA 100 USD
101932-968 101932-970
3-Fluoro-5-formylbenzonitrile ≥95%
3-Fluoro-5-formylbenzonitrile

Hazard: Irritant

Formula: C8H4FNO
Melting Pt: 82…84 °C
MDL Number: MFCD04974123
CAS Number: 1003708-42-6

Order Now

Specification Test Results

Melting point 82-84°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR