Ethyl-3-(dimethylamino)-2-methanesulfonylprop-2-enoate ≥95%

Supplier: Matrix Scientific

067011-500MG 067011-1G
101932-960EA 298.46 USD
101932-960 101932-962
Ethyl-3-(dimethylamino)-2-methanesulfonylprop-2-enoate ≥95%
Ethyl-3-(dimethylamino)-2-methanesulfonylprop-2-enoate

Hazard: Irritant

Formula: C₈H₁₅NO₄S
MW: 221.28 g/mol
Melting Pt: 57…59 °C
MDL Number: MFCD20444438
CAS Number: 1240263-70-0

Order Now

Specification Test Results

Melting point 57-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR