Ethyl-3-cyano-4-(dimethylamino)-2-oxobut-3-enoate ≥95%

Supplier: Matrix Scientific

067010-500MG 067010-1G 067010-5G
101932-954EA 298.46 USD
101932-954 101932-956 101932-958
Ethyl-3-cyano-4-(dimethylamino)-2-oxobut-3-enoate ≥95%
Ethyl-3-cyano-4-(dimethylamino)-2-oxobut-3-enoate

Hazard: Irritant

Formula: C₉H₁₂N₂O₃
MW: 196.21 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD00664335
CAS Number: 339097-11-9

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR