N,N-Dimethyl-1H-pyrrole-2-carboxamide ≥95%

Supplier: Matrix Scientific

067009-500MG 067009-1G
101932-950EA 403.08 USD
101932-950 101932-952
N,N-Dimethyl-1H-pyrrole-2-carboxamide ≥95%
N,N-Dimethyl-1H-pyrrole-2-carboxamide

Hazard: Irritant

Formula: C₇H₁₀N₂O
Melting Pt: 97…99 °C
MDL Number: MFCD12923836
CAS Number: 7126-47-8

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR