4-Chloro-1,6-naphthyridine ≥95%

Supplier: Matrix Scientific

067002-500MG 067002-1G
101932-928EA 298.46 USD
101932-928 101932-930
4-Chloro-1,6-naphthyridine ≥95%
4-Chloro-1,6-naphthyridine

Hazard: Irritant

Formula: C₈H₅ClN₂
Melting Pt: 96…98 °C
MDL Number: MFCD09881194
CAS Number: 6861-84-3

Order Now

Specification Test Results

Melting point 90°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR