5-Chloro-3-hydroxy-2,3-dihydro-benzothiophene-1,1-dione ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2,3-dihydro-1-benzothiophene-3-ol 1,1-dioxide

067001-500MG 067001-1G
101932-924EA 403.08 USD
101932-924 101932-926
5-Chloro-3-hydroxy-2,3-dihydro-benzothiophene-1,1-dione ≥95%
5-Chloro-3-hydroxy-2,3-dihydro-benzothiophene-1,1-dione

Hazard: Irritant

Formula: C₈H₇ClO₃S
MW: 218.66 g/mol
Melting Pt: 136…138 °C
MDL Number: MFCD20444433
CAS Number: 1353878-07-5

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR