2-Bromoquinoxaline ≥95%

Supplier: Matrix Scientific

066999-500MG 066999-1G
101932-916EA 298.46 USD
101932-916 101932-918
2-Bromoquinoxaline ≥95%
2-Bromoquinoxaline

Hazard: Irritant

Formula: C₈H₅BrN₂
Melting Pt: 60…61 °C
MDL Number: MFCD07368206
CAS Number: 36856-91-4

Order Now

Specification Test Results

Melting point 60-61°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR