5-Bromo-2-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

066998-1G 066998-5G
101932-912EA 20 USD
101932-912 101932-914
5-Bromo-2-nitrobenzaldehyde ≥95%
5-Bromo-2-nitrobenzaldehyde

Hazard: Irritant

Formula: C₇H₄BrNO₃
Melting Pt: 70-71°C
MDL Number: MFCD00456508
CAS Number: 20357-20-4

Order Now

Specification Test Results

Melting point 70-71°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR