(5-Bromo-3-iodo-2-methoxyphenyl)methanol ≥95%

Supplier: Matrix Scientific

066996-500MG 066996-1G
101932-904EA 803.08 USD
101932-904 101932-906
(5-Bromo-3-iodo-2-methoxyphenyl)methanol ≥95%
(5-Bromo-3-iodo-2-methoxyphenyl)methanol

Hazard: Irritant

Formula: C₈H₈BrIO₂
Melting Pt: 71…74 °C
MDL Number: MFCD20444430
CAS Number: 1353878-13-3

Order Now

Specification Test Results

Melting point 71-74°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR