5-Bromo-3-iodo-2-methoxybenzaldehyde ≥95%

Supplier: Matrix Scientific

066995-500MG 066995-1G
101932-900EA 995.38 USD
101932-900 101932-902
5-Bromo-3-iodo-2-methoxybenzaldehyde ≥95%
5-Bromo-3-iodo-2-methoxybenzaldehyde

Hazard: Irritant

Formula: C₈H₆BrIO₂
Melting Pt: 96…98 °C
MDL Number: MFCD02257474
CAS Number: 832674-07-4

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR