2-(1-Benzylpiperidin-4-ylidene)acetonitrile ≥95%

Supplier: Matrix Scientific

066993-500MG 066993-1G 066993-5G
101932-890EA 193.85 USD
101932-890 101932-892 101932-894
2-(1-Benzylpiperidin-4-ylidene)acetonitrile ≥95%
2-(1-Benzylpiperidin-4-ylidene)acetonitrile

Hazard: Irritant

Formula: C₁₄H₁₆N₂
Melting Pt: 93…94 °C
MDL Number: MFCD09908274
CAS Number: 55022-82-7

Order Now

Specification Test Results

Melting point 93-94°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR