S-(2-Carboxyethyl)-L-cysteine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-[(2-Carboxyethyl)thio]alanine
066988-500MG
066988-1G
066988-5G
101932-870EA
193.85
USD
101932-870
101932-872
101932-874
S-(2-Carboxyethyl)-L-cysteine ≥95%
S-(2-Carboxyethyl)-L-cysteine
Hazard: Irritant
Formula:
C₆H₁₁NO₄S MW: 193.22 g/mol Melting Pt: 218 °C (dec.) |
MDL Number:
MFCD03424208 CAS Number: 4033-46-9 |
Specification Test Results
Melting point | 218°C(dec) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...