tert-Butyl-3-(trifluoromethyl)-1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4,5,6,7-Tetrahydro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine, N5-BOC protected, tert-Butyl 3-(trifluoromethyl)-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate

066936-500MG 066936-1G
101932-814EA 650.77 USD
101932-814 101932-816
tert-Butyl-3-(trifluoromethyl)-1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate ≥95%
tert-Butyl-3-(trifluoromethyl)-1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₆F₃N₃O₂
MW: 291.28 g/mol
Melting Pt: 170…172 °C
MDL Number: MFCD11044975
CAS Number: 1022931-73-2

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR