Methyl-4-(thiomorpholin-4-ylmethyl)benzoate ≥95%

Supplier: Matrix Scientific

066930-500MG 066930-1G
101932-792EA 298.46 USD
101932-792 101932-794
Methyl-4-(thiomorpholin-4-ylmethyl)benzoate ≥95%
Methyl-4-(thiomorpholin-4-ylmethyl)benzoate

Hazard: Irritant

Formula: C₁₃H₁₇NO₂S
MW: 251.35 g/mol
Melting Pt: 73…75 °C
MDL Number: MFCD01452463
CAS Number: 128982-45-6

Order Now

Specification Test Results

Melting point 73-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR