Ethyl-6-bromo-2-hydrazinyl-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

066924-500MG 066924-1G
101932-768EA 995.38 USD
101932-768 101932-770
Ethyl-6-bromo-2-hydrazinyl-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-6-bromo-2-hydrazinyl-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁BrN₄O₂
MW: 311.14 g/mol
Melting Pt: 180…182 °C
MDL Number: MFCD20232917
CAS Number: 1335113-09-1

Order Now

Specification Test Results

Melting point 180-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR