4-Chlorothiazole-5-carboxaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chlorothiazole-5-carbaldehyde

066921-500MG 066921-1G
101932-760EA 90.77 USD
101932-760 101932-762
4-Chlorothiazole-5-carboxaldehyde ≥95%
4-Chlorothiazole-5-carboxaldehyde

Hazard: Irritant

Formula: C₄H₂ClNOS
MW: 147.5828 g/mol
Melting Pt: 53…54 °C
MDL Number: MFCD09837291
CAS Number: 104146-17-0

Order Now

Specification Test Results

Melting point 53-54°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR