6-Chloro-1H-pyrazolo[3,4-b]1,8-naphthyridin-3-amine ≥95%
Supplier: Matrix Scientific
066920-500MG
066920-1G
101932-756EA
1192.31
USD
101932-756
101932-758
6-Chloro-1H-pyrazolo[3,4-b]1,8-naphthyridin-3-amine ≥95%
6-Chloro-1H-pyrazolo[3,4-b]1,8-naphthyridin-3-amine
Hazard: Irritant
Formula:
C₉H₆ClN₅ Melting Pt: >300 °C |
MDL Number:
MFCD20232918 CAS Number: 1335220-58-0 |
Specification Test Results
Melting point | >300°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...