6-Chloro-2-(piperidin-1-yl)-1,8-naphthyridine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

066919-500MG 066919-1G
101932-752EA 1192.31 USD
101932-752 101932-754
6-Chloro-2-(piperidin-1-yl)-1,8-naphthyridine-3-carbonitrile ≥95%
6-Chloro-2-(piperidin-1-yl)-1,8-naphthyridine-3-carbonitrile

Hazard: Irritant

Formula: C₁₄H₁₃ClN₄
Melting Pt: 146…148 °C
MDL Number: MFCD20232923
CAS Number: 1335220-59-1

Order Now

Specification Test Results

Melting point 146-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR