1-Bromo-3-methoxynaphthalen-2-ol ≥95%

Supplier: Matrix Scientific

066913-500MG 066913-1G
101932-736EA 600 USD
101932-736 101932-738
1-Bromo-3-methoxynaphthalen-2-ol ≥95%
1-Bromo-3-methoxynaphthalen-2-ol

Hazard: Irritant

Formula: C₁₁H₉BrO₂
Melting Pt: 98…99 °C
MDL Number: MFCD20232913
CAS Number: 404965-07-7

Order Now

Specification Test Results

Melting point 98-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR