1-(5-Bromo-6-methoxy-2-methylpyridin-3-yl)ethan-1-one ≥95%

Supplier: Matrix Scientific

066912-500MG 066912-1G
101932-732EA 995.38 USD
101932-732 101932-734
1-(5-Bromo-6-methoxy-2-methylpyridin-3-yl)ethan-1-one ≥95%
1-(5-Bromo-6-methoxy-2-methylpyridin-3-yl)ethan-1-one

Hazard: Irritant

Formula: C₉H₁₀BrNO₂
Melting Pt: 89…91 °C
MDL Number: MFCD20232925
CAS Number: 1335113-01-3

Order Now

Specification Test Results

Melting point 89-91°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR