2,4,6-Tris(methylsulfanyl)-1,3,5-triazine ≥95%

Supplier: Matrix Scientific

066900-500MG 066900-1G 066900-5G
101932-694EA 403.08 USD
101932-694 101932-696 101932-698
2,4,6-Tris(methylsulfanyl)-1,3,5-triazine ≥95%
2,4,6-Tris(methylsulfanyl)-1,3,5-triazine

Hazard: Irritant

Formula: C₆H₉N₃S₃
Melting Pt: 188…190 °C
MDL Number: MFCD01678800
CAS Number: 5759-58-0

Order Now

Specification Test Results

Melting point 188-190°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR