1-[3-(Trifluoromethyl)pyridin-2-yl]ethan-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Acetyl-3-(trifluoromethyl)pyridine

066899-500MG 066899-1G
101932-690EA 638.46 USD
101932-690 101932-692
1-[3-(Trifluoromethyl)pyridin-2-yl]ethan-1-one ≥95%
1-[3-(Trifluoromethyl)pyridin-2-yl]ethan-1-one

Hazard: Irritant

Formula: C₈H₆F₃NO
MW: 189.14 g/mol
Melting Pt: 65…70 °C
MDL Number: MFCD11044107
CAS Number: 749257-78-1

Order Now

Specification Test Results

Melting point 65-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR