1-{2-[4-(Trifluoromethyl)piperidin-1-yl]ethyl}imidazolidin-2-one ≥95%

Supplier: Matrix Scientific

066898-500MG 066898-1G
101932-686EA 1192.31 USD
101932-686 101932-688
1-{2-[4-(Trifluoromethyl)piperidin-1-yl]ethyl}imidazolidin-2-one ≥95%
1-{2-[4-(Trifluoromethyl)piperidin-1-yl]ethyl}imidazolidin-2-one

Hazard: Irritant

Formula: C₁₁H₁₈F₃N₃O
Melting Pt: 126…127 °C
MDL Number: MFCD19982784
CAS Number: 1191047-90-1

Order Now

Specification Test Results

Melting point 126-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR