1-tert-Butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine ≥95%
Supplier: Matrix Scientific
066896-500MG
066896-1G
101932-678EA
600
USD
101932-678
101932-680
1-tert-Butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine ≥95%
1-tert-Butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Hazard: Irritant
Formula:
C₉H₁₃N₅ Melting Pt: 190…192 °C |
MDL Number:
MFCD00128294 CAS Number: 862728-60-7 |
Specification Test Results
Melting point | 190-192°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...