tert-Butyl-4-(cyanomethylidene)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(cyanomethylene)piperidine-1-carboxylate

066893-500MG 066893-1G 066893-5G
101932-662EA 138.46 USD
101932-662 101932-664 101932-666
tert-Butyl-4-(cyanomethylidene)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(cyanomethylidene)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
Melting Pt: 121…122 °C
MDL Number: MFCD03695496
CAS Number: 197569-11-2

Order Now

Specification Test Results

Melting point 121-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR