3-Phenyl-1-oxa-3,8-diazaspiro[4.5]decan-2-one ≥95%

Supplier: Matrix Scientific

066891-500MG 066891-1G
101932-656EA 1192.31 USD
101932-656 101932-658
3-Phenyl-1-oxa-3,8-diazaspiro[4.5]decan-2-one ≥95%
3-Phenyl-1-oxa-3,8-diazaspiro[4.5]decan-2-one

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₂
Melting Pt: 158…160 °C
MDL Number: MFCD19982780
CAS Number: 77211-58-6

Order Now

Specification Test Results

Melting point 158-160°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR