4-{[(4-Fluorophenyl)amino]methyl}piperidin-4-ol ≥95%

Supplier: Matrix Scientific

066887-500MG 066887-1G
101932-638EA 803.08 USD
101932-638 101932-640
4-{[(4-Fluorophenyl)amino]methyl}piperidin-4-ol ≥95%
4-{[(4-Fluorophenyl)amino]methyl}piperidin-4-ol

Hazard: Irritant

Formula: C₁₂H₁₇FN₂O
Melting Pt: 141…143 °C
MDL Number: MFCD19982788
CAS Number: 1330583-68-0

Order Now

Specification Test Results

Melting point 141-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR