Ethyl-7-chloro-8-iodoimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

066884-500MG 066884-1G
101932-626EA 704.62 USD
101932-626 101932-628
Ethyl-7-chloro-8-iodoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl-7-chloro-8-iodoimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈ClIN₂O₂
Melting Pt: 217…218 °C
MDL Number: MFCD19982795
CAS Number: 1331823-97-2

Order Now

Specification Test Results

Melting point 217-218°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR