Ethyl-6-chloro-2-hydroxy-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

066883-500MG 066883-1G
101932-622EA 707.69 USD
101932-622 101932-624
Ethyl-6-chloro-2-hydroxy-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-6-chloro-2-hydroxy-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₃
Melting Pt: 252…254 °C
MDL Number: MFCD19982793
CAS Number: 1330583-64-6

Order Now

Specification Test Results

Melting point 252-254°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR