Ethyl-6-bromo-2-chloro-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

066882-500MG 066882-1G
101932-618EA 803.08 USD
101932-618 101932-620
Ethyl-6-bromo-2-chloro-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-6-bromo-2-chloro-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₈BrClN₂O₂
Melting Pt: 144…146 °C
MDL Number: MFCD19982791
CAS Number: 1330583-62-4

Order Now

Specification Test Results

Melting point 144-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR