5,6-Dichloronicotinamide ≥95%

Supplier: Matrix Scientific

066881-500MG 066881-1G 066881-5G
101932-612EA 230.77 USD
101932-612 101932-614 101932-616
5,6-Dichloronicotinamide ≥95%
5,6-Dichloronicotinamide

Hazard: Irritant

Formula: C₆H₄Cl₂N₂O
Melting Pt: 223-225°C
CAS Number: 75291-84-8

Order Now

Specification Test Results

Melting point 223-225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR