cis-3-[(tert-Butoxy)carbonyl]-3-azabicyclo[3.1.0]hexane-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

066878-500MG 066878-1G
101932-600EA 1524.62 USD
101932-600 101932-602
cis-3-[(tert-Butoxy)carbonyl]-3-azabicyclo[3.1.0]hexane-2-carboxylic acid ≥95%
cis-3-[(tert-Butoxy)carbonyl]-3-azabicyclo[3.1.0]hexane-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₇NO₄
Melting Pt: 133…134 °C
MDL Number: MFCD10676737
CAS Number: 72448-25-0

Order Now

Specification Test Results

Melting point 133-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR