5-Chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

066877-500MG 066877-1G
101932-596EA 803.08 USD
101932-596 101932-598
5-Chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid ≥95%
5-Chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₅ClN₂O₂
Melting Pt: 300 °C+
MDL Number: MFCD18261123
CAS Number: 800401-63-2

Order Now

Specification Test Results

Melting point 300°C+

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR