4-Chloro-5-methoxybenzene-1,2-diamine ≥95%

Supplier: Matrix Scientific

066874-500MG 066874-1G
101932-584EA 453.85 USD
101932-584 101932-586
4-Chloro-5-methoxybenzene-1,2-diamine ≥95%
4-Chloro-5-methoxybenzene-1,2-diamine

Hazard: Irritant

Formula: C₇H₉ClN₂O
Melting Pt: 127…129 °C
MDL Number: MFCD19982792
CAS Number: 38608-06-9

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR