7-Chloro-8-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
Supplier: Matrix Scientific
066873-500MG
066873-1G
101932-580EA
803.08
USD
101932-580
101932-582
7-Chloro-8-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
7-Chloro-8-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₈H₃ClF₃IN₂ MW: 346.48 g/mol Melting Pt: 160…162 °C |
MDL Number:
MFCD19982794 CAS Number: 1331823-98-3 |
Specification Test Results
Melting point | 160-162°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...