3-Bromo-5-chloro-1H-indazole ≥95%

Supplier: Matrix Scientific

066869-500MG 066869-1G
101932-564EA 600 USD
101932-564 101932-566
3-Bromo-5-chloro-1H-indazole ≥95%
3-Bromo-5-chloro-1H-indazole

Hazard: Irritant

Formula: C₇H₄BrClN₂
Melting Pt: 233…235 °C
MDL Number: MFCD07781584
CAS Number: 885521-43-7

Order Now

Specification Test Results

Melting point 233-235°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR