7-Nitro-2(1H)-quinoxalinone ≥95%

Supplier: Matrix Scientific

066807-500MG 066807-1G
101932-430EA 600 USD
101932-430 101932-432
7-Nitro-2(1H)-quinoxalinone ≥95%
7-Nitro-2(1H)-quinoxalinone

Hazard: Irritant

Formula: C₈H₅N₃O₃
MW: 191.1436 g/mol
Melting Pt: 281…282 °C
MDL Number: MFCD09999191
CAS Number: 89898-96-4

Order Now

Specification Test Results

Melting point 281-282°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR