Methyl-2-(6-cyano-2H-1,3-benzodioxol-5-yl)acetate ≥95%

Supplier: Matrix Scientific

066805-500MG 066805-1G
101932-422EA 600 USD
101932-422 101932-424
Methyl-2-(6-cyano-2H-1,3-benzodioxol-5-yl)acetate ≥95%
Methyl-2-(6-cyano-2H-1,3-benzodioxol-5-yl)acetate

Hazard: Irritant

Formula: C₁₁H₉NO₄
Melting Pt: 87 °C (subl.)
MDL Number: MFCD19686129
CAS Number: 184042-03-3

Order Now

Specification Test Results

Melting point 87°C(sub)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR