3-Iodo-1-methyl-5-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

066804-500MG 066804-1G
101932-418EA 995.38 USD
101932-418 101932-420
3-Iodo-1-methyl-5-nitro-1H-indazole ≥95%
3-Iodo-1-methyl-5-nitro-1H-indazole

Hazard: Irritant

Formula: C₈H₆IN₃O₂
Melting Pt: 199…201 °C
MDL Number: MFCD19686128
CAS Number: 1315577-17-3

Order Now

Specification Test Results

Melting point 199-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR