(2-Fluoro-4-nitrophenyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-4-nitrobenzyl alcohol

066800-500MG 066800-1G 066800-5G
101932-400EA 156.92 USD
101932-400 101932-402 101932-404
(2-Fluoro-4-nitrophenyl)methanol ≥95%
(2-Fluoro-4-nitrophenyl)methanol

Hazard: Irritant

Formula: C₇H₆FNO₃
Melting Pt: 103…105 °C
MDL Number: MFCD11110168
CAS Number: 660432-43-9

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR