Ethyl-6-chloro-4-hydrazinylpyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

066796-500MG 066796-1G
101932-382EA 600 USD
101932-382 101932-384
Ethyl-6-chloro-4-hydrazinylpyridine-3-carboxylate ≥95%
Ethyl-6-chloro-4-hydrazinylpyridine-3-carboxylate

Hazard: Irritant

Formula: C₈H₁₀ClN₃O₂
MW: 215.64 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD19686131
CAS Number: 845655-96-1

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR