2-(6-Cyano-2H-1,3-benzodioxol-5-yl)acetic acid ≥95%

Supplier: Matrix Scientific

066794-500MG 066794-1G
101932-374EA 803.08 USD
101932-374 101932-376
2-(6-Cyano-2H-1,3-benzodioxol-5-yl)acetic acid ≥95%
2-(6-Cyano-2H-1,3-benzodioxol-5-yl)acetic acid

Hazard: Irritant

Formula: C₁₀H₇NO₄
Melting Pt: 230 °C (dec.)
MDL Number: MFCD19686130
CAS Number: 1315577-19-5

Order Now

Specification Test Results

Melting point 230°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR