1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]azetidin-3-ol ≥95%

Supplier: Matrix Scientific

066791-500MG 066791-1G
101932-364EA 403.08 USD
101932-364 101932-366
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]azetidin-3-ol ≥95%
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]azetidin-3-ol

Hazard: Irritant

Formula: C₉H₈ClF₃N₂O
Melting Pt: 88…90 °C
MDL Number: MFCD18266134
CAS Number: 1311798-09-0

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR