2-Chloro-7-nitroquinoxaline ≥95%

Supplier: Matrix Scientific

066789-500MG 066789-1G
101932-356EA 803.08 USD
101932-356 101932-358
2-Chloro-7-nitroquinoxaline ≥95%
2-Chloro-7-nitroquinoxaline

Hazard: Irritant

Formula: C₈H₄ClN₃O₂
MW: 209.59 g/mol
Melting Pt: 185…187 °C
MDL Number: MFCD00033669
CAS Number: 55686-94-7

Order Now

Specification Test Results

Melting point 185-187°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR