6-Bromooxazolo[4,5-b]pyridine ≥95%

Supplier: Matrix Scientific

066787-500MG 066787-1G
101932-348EA 215.38 USD
101932-348 101932-350
6-Bromooxazolo[4,5-b]pyridine ≥95%
6-Bromooxazolo[4,5-b]pyridine

Hazard: Irritant

Formula: C₆H₃BrN₂O
Melting Pt: 156…158 °C
MDL Number: MFCD13193309
CAS Number: 1260863-86-2

Order Now

Specification Test Results

Melting point 156-158°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR