1-Benzyl-1H,4H,5H,6H-cyclopenta[b]pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

066783-500MG 066783-1G
101932-330EA 403.08 USD
101932-330 101932-332
1-Benzyl-1H,4H,5H,6H-cyclopenta[b]pyrrole-2-carboxylic acid ≥95%
1-Benzyl-1H,4H,5H,6H-cyclopenta[b]pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₅NO₂
Melting Pt: 163…164 °C
MDL Number: MFCD19686124
CAS Number: 164736-53-2

Order Now

Specification Test Results

Melting point 163-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR