N-(2-Amino-5-chloro-4-methoxyphenyl)acetamide ≥95%

Supplier: Matrix Scientific

066782-500MG 066782-1G
101932-326EA 486.15 USD
101932-326 101932-328
N-(2-Amino-5-chloro-4-methoxyphenyl)acetamide ≥95%
N-(2-Amino-5-chloro-4-methoxyphenyl)acetamide

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂
Melting Pt: 150…152 °C
MDL Number: MFCD19686132
CAS Number: 857553-74-3

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR