6-Bromoanthranilic acid ≥96%

Supplier: Matrix Scientific

066628-1G 066628-5G
101932-014EA 23.08 USD
101932-014 101932-016
6-Bromoanthranilic acid ≥96%
6-Bromoanthranilic acid

Hazard: Irritant

Formula: C₇H₆BrNO₂
Melting Pt: 134…135 °C
MDL Number: MFCD03618455
CAS Number: 20776-48-1

Order Now

Specification Test Results

Melting point 134-135°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR