Methyl-6-chloro-4-methoxypyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-chloro-4-methoxynicotinate

064091-500MG 064091-1G
101927-028EA 500 USD
101927-028 101927-030
Methyl-6-chloro-4-methoxypyridine-3-carboxylate ≥95%
Methyl-6-chloro-4-methoxypyridine-3-carboxylate

Hazard: Irritant

Formula: C₈H₈ClNO₃
Melting Pt: 73…75 °C
MDL Number: MFCD12198235
CAS Number: 84332-02-5

Order Now

Specification Test Results

Melting point 73-75°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR