Methyl-6-chloro-2-methoxypyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-chloro-2-methoxynicotinate

064090-500MG 064090-1G
101927-024EA 600 USD
101927-024 101927-026
Methyl-6-chloro-2-methoxypyridine-3-carboxylate ≥95%
Methyl-6-chloro-2-methoxypyridine-3-carboxylate

Hazard: Irritant

Formula: C₈H₈ClNO₃
Melting Pt: 56…57 °C
MDL Number: MFCD15142741
CAS Number: 65515-32-4

Order Now

Specification Test Results

Melting point 56-57°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR